(3,5-Difluor-4-methoxyphenyl)essigsäure 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3,5-Difluoro-4-methoxyphenylacetic acid
PC6037-1G
PC6037-5G
PC6037-10G
PC6037-25G
APOSPC6037-1GEA
142
EUR
APOSPC6037-1G
APOSPC6037-5G
APOSPC6037-10G
APOSPC6037-25G
(3,5-Difluor-4-methoxyphenyl)essigsäure 98%
(3,5-Difluor-4-methoxyphenyl)essigsäure
Formel:
C₉H₈F₂O₃ Molecular Weight: 202,15 g/mol Schmelzpunkt: 85…86 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD04115919 CAS-Nummer: 886498-74-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...