Ethyl-4-(trifluoromethyl)-1-benzothiophene-2-carboxylate

Lieferant: Apollo Scientific
Achtung

PC6051-1G PC6051-5G
APOSPC6051-1GEA 125 EUR
APOSPC6051-1G APOSPC6051-5G
Ethyl-4-(trifluoromethyl)-1-benzothiophene-2-carboxylate
Ethyl-4-(trifluoromethyl)-1-benzothiophene-2-carboxylate
Formel: C₁₂H₉F₃O₂S
Lagertemperatur: Kühlschrank
MDL: MFCD11100109
CAS-Nummer: 1209178-96-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR