4-Chlor-2-iod-5-(trifluormethyl)anilin

Lieferant: Apollo Scientific

Synonyme: 5-Amino-2-chloro-4-iodobenzotrifluoride

PC6061-1G PC6061-500MG PC6061-5G
APOSPC6061-1GEA 219 EUR
APOSPC6061-1G APOSPC6061-500MG APOSPC6061-5G
4-Chlor-2-iod-5-(trifluormethyl)anilin
4-Chlor-2-iod-5-(trifluormethyl)anilin
Formel: C₇H₄ClF₃IN
Molecular Weight: 321,466 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD12827773
CAS-Nummer: 852569-36-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR