3-Formyl-6-(trifluoromethyl)-1H-indole-5-carbonitrile

Lieferant: Apollo Scientific

PC6063-1G PC6063-500MG
APOSPC6063-1GEA 539 EUR
APOSPC6063-1G APOSPC6063-500MG
3-Formyl-6-(trifluoromethyl)-1H-indole-5-carbonitrile
3-Formyl-6-(trifluoromethyl)-1H-indole-5-carbonitrile
Formel: C₁₁H₅F₃N₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD12827826
CAS-Nummer: 1186404-95-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR