5-Chloro-3-fluoropyridine-2-carboxylic acid

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Chloro-3-fluoro-2-pyridinecarboxylic acid, 5-Chloro-3-fluoropicolinic acid

PC6073-5G PC6073-1G PC6073-250MG
APOSPC6073-5GEA 578 EUR
APOSPC6073-5G APOSPC6073-1G APOSPC6073-250MG
5-Chloro-3-fluoropyridine-2-carboxylic acid
5-Chloro-3-fluoropyridine-2-carboxylic acid
Formel: C₆H₃ClFNO₂
Molecular Weight: 175.55 g/mol
Schmelzpunkt: 172…174 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD12827555
CAS-Nummer: 207994-08-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR