2,5-Dibrom-3-fluorpyridin 98%

Lieferant: Apollo Scientific
Achtung

PC6077-5G PC6077-10G PC6077-1G
APOSPC6077-5GEA 87 EUR
APOSPC6077-5G APOSPC6077-10G APOSPC6077-1G
2,5-Dibrom-3-fluorpyridin 98%
2,5-Dibromo-3-fluoropyridine
Formel: C₅H₂Br₂FN
Molecular Weight: 254,88 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06659497
CAS-Nummer: 156772-60-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR