4-Amino-3-fluorphenol Hydrochlorid 95%

Lieferant: Apollo Scientific
Achtung

PC6096-25G
APOSPC6096-25GEA 102 EUR
APOSPC6096-25G
4-Amino-3-fluorphenol Hydrochlorid 95%
4-Amino-3-fluorphenol Hydrochlorid
Formel: C₆H₇ClFNO
Molecular Weight: 163,5773 g/mol
Flammpunkt: >110 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01632189
CAS-Nummer: 18266-53-0
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR