1H,1H,2H,2H-Perfluoroctyltriethoxysilan 98%
Lieferant: Apollo Scientific
Achtung
Synonyme: Triethoxy-1H,1H,2H,2H-tridecafluoro-n-octylsilane, Tridecafluoro-1,1,2,2-tetrahydrooctyl-1-triethoxysilane, Triethoxy(3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctyl)silane;
PC6181-10G PC6181-100G PC6181-25G
APOSPC6181-10GEA 21.9 EUR
APOSPC6181-10G APOSPC6181-100G APOSPC6181-25G
1H,1H,2H,2H-Perfluoroctyltriethoxysilan 98%
1H,1H,2H,2H-Perfluoroctyltriethoxysilan
1H,1H,2H,2H-perfluorooctyltriethoxysilane (CAS 51851-37-7) is a flourinated alkyl silane. Apollo Scientific supplies 1H,1H,2H,2H-perfluorooctyltriethoxysilane to R&D, Academic institutions, and manufacturers across the UK and USA.
It is commonly used as a surface modifier or surface treatment agent to render surfaces superhydrophobic, such as titanium or carbon nanotubes (CNT) hollow membranes.
Poly(vinylidene flouride) (PVDF) may also be coated with 1H,1H,2H,2H-perfluorooctyltriethoxysilane to synthesise a membrane to remove organic compounds which may be hazardous and volatile. It can also be used in the silanisation of silicon wafer, or the production of chemical surface patterns of self assembled monolayers.
Formel: C₁₄H₁₉F₁₃O₃Si
Molecular Weight: 510,367 g/mol
Siedepunkt: 84…86 °C/1,5mmHg
Dichte: 1,3299 g/cm³ (25 °C)
Flammpunkt: 97 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00042333
CAS-Nummer: 51851-37-7
EINECS: 257-473-3