3-Brom-5-methylbenzotrifluorid 99%

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-Bromo-3-methyl-5-(trifluoromethyl)benzene

PC6188-10G PC6188-5G PC6188-1G
APOSPC6188-10GEA 60.9 EUR
APOSPC6188-10G APOSPC6188-5G APOSPC6188-1G
3-Brom-5-methylbenzotrifluorid 99%
3-Brom-5-methylbenzotrifluorid
Formel: C₈H₆BrF₃
Molecular Weight: 239,04 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01631585
CAS-Nummer: 86845-28-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR