2,5-Dichloro-4-(trifluoromethyl)pyrimidine

Lieferant: Apollo Scientific

PC6207-100MG PC6207-250MG
APOSPC6207-100MGEA 29 EUR
APOSPC6207-100MG APOSPC6207-250MG
2,5-Dichloro-4-(trifluoromethyl)pyrimidine
2,5-Dichloro-4-(trifluoromethyl)pyrimidine
Formel: C₅HCl₂F₃N₂
Molecular Weight: 216.98 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13176599
CAS-Nummer: 785777-98-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR