2'-Chlor-4'-(trifluormethoxy)acetanilid
Lieferant: Apollo Scientific
|
Achtung
|
PC6218-1G
PC6218-5G
PC6218-25G
APOSPC6218-1GEA
29
EUR
APOSPC6218-1G
APOSPC6218-5G
APOSPC6218-25G
2'-Chlor-4'-(trifluormethoxy)acetanilid
2'-Chlor-4'-(trifluormethoxy)acetanilid
Formel:
C₉H₇ClF₃NO₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD01571100 CAS-Nummer: 256529-43-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...