8-(Trifluormethyl)-4-chinolinol
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-Hydroxy-8-(trifluoromethyl)quinoline
PC6241-1G
PC6241-5G
APOSPC6241-1GEA
23.2
EUR
APOSPC6241-1G
APOSPC6241-5G
8-(Trifluormethyl)-4-chinolinol
8-(Trifluormethyl)-4-chinolinol
Formel:
C₁₀H₆F₃NO Molecular Weight: 213,16 g/mol Schmelzpunkt: 174…176 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00134577 CAS-Nummer: 23779-96-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...