2,5-Dichloro-3-fluoropyridine 96%
Lieferant: Apollo Scientific
|
Achtung
|
PC6247-1G
PC6247-5G
PC6247-25G
APOSPC6247-1GEA
72.5
EUR
APOSPC6247-1G
APOSPC6247-5G
APOSPC6247-25G
2,5-Dichloro-3-fluoropyridine 96%
2,5-Dichloro-3-fluoropyridine
Formel:
C₅H₂Cl₂FN Schmelzpunkt: 54…56 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD10699709 CAS-Nummer: 103999-77-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...