5-Chloro-2,4-difluoroaniline 97%

Lieferant: Apollo Scientific

PC6270-1G PC6270-5G PC6270-10G PC6270-25G
APOSPC6270-1GEA 17.6 EUR
APOSPC6270-1G APOSPC6270-5G APOSPC6270-10G APOSPC6270-25G
5-Chloro-2,4-difluoroaniline 97%
5-Chloro-2,4-difluoroaniline
Formel: C6H4ClF2N
Molecular Weight: 163.5525 g/mol
Schmelzpunkt: 50…53 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD06808806
CAS-Nummer: 348-65-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR