2-Amino-2-[3-(trifluoromethyl)phenyl]acetic acid 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-(Trifluoromethyl)phenylglycine, H-DL-Phg(3-CF₃)-OH, 2-Amino-2-(3-(trifluoromethyl)phenyl)acetic acid

PC6289-100MG PC6289-500MG
APOSPC6289-100MGEA 58.2 EUR
APOSPC6289-100MG APOSPC6289-500MG
2-Amino-2-[3-(trifluoromethyl)phenyl]acetic acid 97%
2-Amino-2-[3-(trifluoromethyl)phenyl]acetic acid
Formel: C₉H₈F₃NO₂
Molecular Weight: 219,16 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02662404
CAS-Nummer: 242475-26-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR