3-Cyan-4-fluorbenzolsulfonylchlorid 97%

Lieferant: Apollo Scientific
Gefahr

PC6312-5G PC6312-25G
APOSPC6312-5GEA 46.4 EUR
APOSPC6312-5G APOSPC6312-25G
3-Cyan-4-fluorbenzolsulfonylchlorid 97%
3-Cyan-4-fluorbenzolsulfonylchlorid
Formel: C₇H₃ClFNO₂S
Molecular Weight: 219.62 g/mol
Schmelzpunkt: 67…70 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03093760
CAS-Nummer: 351003-23-1
UN: 3261
ADR: 8 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR