5-(4-Chlorophenoxy)-1-methyl-3-(trifluoromethyl)-1H-pyrazole-4-carbaldehyde 97%
Lieferant: Apollo Scientific
|
Achtung
|
PC6326-1G
PC6326-5G
APOSPC6326-1GEA
302
EUR
APOSPC6326-1G
APOSPC6326-5G
5-(4-Chlorophenoxy)-1-methyl-3-(trifluoromethyl)-1H-pyrazole-4-carbaldehyde 97%
5-(4-Chlorophenoxy)-1-methyl-3-(trifluoromethyl)-1H-pyrazole-4-carbaldehyde
Formel:
C₁₂H₈ClF₃N₂O₂ Molecular Weight: 304,66 g/mol Schmelzpunkt: 125…126 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01935951 CAS-Nummer: 321848-47-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...