5-Chlor-2-(trifluormethyl)benzoesäure 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
5-Chlor-α,α,α-trifluor-o-toluylsäure
PC6347-25G
PC6347-5G
PC6347-1G
APOSPC6347-25GEA
191
EUR
APOSPC6347-25G
APOSPC6347-5G
APOSPC6347-1G
5-Chlor-2-(trifluormethyl)benzoesäure 98%
5-Chlor-2-(trifluormethyl)benzoesäure
Formel:
C₈H₄ClF₃O₂ Molecular Weight: 224.57 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD01631353 CAS-Nummer: 654-98-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...