2-Nitro-5-(trifluoromethyl)benzenol
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Nitro-5-(trifluoromethyl)phenol, 3-Hydroxy-4-nitrobenzotrifluoride
PC6361-1G
PC6361-5G
PC6361-25G
APOSPC6361-1GEA
29
EUR
APOSPC6361-1G
APOSPC6361-5G
APOSPC6361-25G
2-Nitro-5-(trifluoromethyl)benzenol
2-Nitro-5-(trifluoromethyl)benzenol
Formel:
C₇H₄F₃NO₃ Molecular Weight: 207,11 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD08282785 CAS-Nummer: 402-17-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...