Silver heptafluorobutyrate 97%
Lieferant: Apollo Scientific
|
Gefahr
|
PC6384-1G
PC6384-25G
PC6384-5G
APOSPC6384-1GEA
0
EUR
APOSPC6384-1G
APOSPC6384-25G
APOSPC6384-5G
Silver heptafluorobutyrate 97%
Silver heptafluorobutyrate
Formel:
C₄AgF₇O₂ Schmelzpunkt: 292…294 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00064197 CAS-Nummer: 3794-64-7 EINECS: 223-266-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...