Silbertetrafluoroborat 99%

Lieferant: Apollo Scientific
Gefahr

Synonyme: Argentous tetrafluoroborate

PC6430-100G PC6430-10G PC6430-50G
APOSPC6430-100GEA 335 EUR
APOSPC6430-100G APOSPC6430-10G APOSPC6430-50G
Silbertetrafluoroborat 99%
Silbertetrafluoroborat

Converts alpha-bromoalkanones to the corresponding fluoro-derivative, Tetrahedron Lett., 1979, 3357.Soluble in many organic solvents.

Formel: AgBF₄
Molecular Weight: 194,67 g/mol
Schmelzpunkt: 70…73 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00003408
CAS-Nummer: 14104-20-2
EINECS: 237-956-5
UN: 3260
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR