2-(1,1,2,2-Tetrafluorethoxy)benzoesäure 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-(2H-Perfluoroethoxy)benzoic acid
PC6499-5G
PC6499-1G
APOSPC6499-5GEA
121
EUR
APOSPC6499-5G
APOSPC6499-1G
2-(1,1,2,2-Tetrafluorethoxy)benzoesäure 98%
2-(1,1,2,2-Tetrafluoroethoxy)benzoic acid
Formel:
C₉H₆F₄O₃ Schmelzpunkt: 78…79 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03094352 CAS-Nummer: 10008-97-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...