(3-Fluor-4-methoxyphenyl)acetonitril 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Fluoro-4-methoxylphenylacetonitrile

PC6574-1G PC6574-5G PC6574-10G
APOSPC6574-1GEA 21.9 EUR
APOSPC6574-1G APOSPC6574-5G APOSPC6574-10G
(3-Fluor-4-methoxyphenyl)acetonitril 98%
(3-Fluor-4-methoxyphenyl)acetonitril
Formel: C₉H₈FNO
Molecular Weight: 165,17 g/mol
Schmelzpunkt: 47…49 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00040889
CAS-Nummer: 404-90-0
UN: 3439
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR