6-Fluor-2-naphthoesäure 97%

Lieferant: Apollo Scientific
Achtung

PC6622-1G PC6622-5G
APOSPC6622-1GEA 132 EUR
APOSPC6622-1G APOSPC6622-5G
6-Fluor-2-naphthoesäure 97%
6-Fluor-2-naphthoesäure
Formel: C₁₁H₇FO₂
Molecular Weight: 190,1700 g/mol
Schmelzpunkt: 242…246 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD06200717
CAS-Nummer: 5043-01-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR