2,2-Difluor-4-pentensäure
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
2,2-Difluoropent-4-enoic acid
PC6655-1G
PC6655-25G
PC6655-5G
APOSPC6655-1GEA
72.5
EUR
APOSPC6655-1G
APOSPC6655-25G
APOSPC6655-5G
2,2-Difluor-4-pentensäure
2,2-Difluoro-4-pentenoic acid
Formel:
C₅H₆F₂O₂ Molecular Weight: 136.1 g/mol Siedepunkt: 73 °C/11mmHg Flammpunkt: 55 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD09800642 CAS-Nummer: 55039-89-9 UN: 3265 ADR: 8 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...