2,3,4-Trifluoriodbenzol
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1,2,3-Trifluoro-4-iodobenzene
PC6691-100G
PC6691-25G
PC6691-5G
APOSPC6691-100GEA
29
EUR
APOSPC6691-100G
APOSPC6691-25G
APOSPC6691-5G
2,3,4-Trifluoriodbenzol
2,3,4-Trifluoriodbenzol
Formel:
C₆H₂F₃I Molecular Weight: 257.9798 g/mol Flammpunkt: 68 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03788551 CAS-Nummer: 459424-72-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...