6-(Trifluormethyl)nicotinsäurehydrazid 95+%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-(Trifluoromethyl)pyridine-5-carboxylic acid hydrazide

PC6698-1G PC6698-5G
APOSPC6698-1GEA 156 EUR
APOSPC6698-1G APOSPC6698-5G
6-(Trifluormethyl)nicotinsäurehydrazid 95+%
6-(Trifluormethyl)nicotinsäurehydrazid
Formel: C₇H₆F₅N₃O
Schmelzpunkt: 128…131 °C
Lagertemperatur: Kühlschrank
MDL: MFCD01862651
CAS-Nummer: 386715-32-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR