tert-Butyl-2-{[2-amino-4-(trifluoromethyl)phenyl]amino}ethylcarbamate 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: tert-Butyl N-{2-[2-amino-4-(trifluoromethyl)anilino]ethyl}carbamate

PC6705-1G PC6705-250MG
APOSPC6705-1GEA 94.4 EUR
APOSPC6705-1G APOSPC6705-250MG
tert-Butyl-2-{[2-amino-4-(trifluoromethyl)phenyl]amino}ethylcarbamate 95%
tert-Butyl-2-{[2-amino-4-(trifluoromethyl)phenyl]amino}ethylcarbamate
Formel: C₁₄H₂₀F₃N₃O₂
Molecular Weight: 319,33 g/mol
Schmelzpunkt: 129…137 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00728760
CAS-Nummer: 215655-42-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR