2-Chlor-3-(trifluormethyl)benzoesäure 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Chlor-α,α,α-trifluor-m-toluylsäure
PC6734-5G
PC6734-1G
PC6734-25G
APOSPC6734-5GEA
24.8
EUR
APOSPC6734-5G
APOSPC6734-1G
APOSPC6734-25G
2-Chlor-3-(trifluormethyl)benzoesäure 98%
2-Chlor-3-(trifluormethyl)benzoesäure
Formel:
C₈H₄ClF₃O₂ Molecular Weight: 224.57 g/mol Schmelzpunkt: 132…134 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00792415 CAS-Nummer: 39226-97-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...