3',4',5'-Trifluoracetophenon 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-(3,4,5-Trifluorophenyl)ethan-1-one

PC6755-5G PC6755-100G PC6755-25G
APOSPC6755-5GEA 21.9 EUR
APOSPC6755-5G APOSPC6755-100G APOSPC6755-25G
3',4',5'-Trifluoracetophenon 98%
3',4',5'-Trifluoracetophenon
Formel: C₈H₅F₃O
Molecular Weight: 174,12 g/mol
Flammpunkt: >100 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00083533
CAS-Nummer: 220141-73-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR