4-Fluor-L-phenylalanin 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(S)-2-Amino-3-(4-fluorophenyl)propanoic acid, H-Phe(4-F)-OH
PC6759-1G
PC6759-5G
PC6759-25G
APOSPC6759-1GEA
21.9
EUR
APOSPC6759-1G
APOSPC6759-5G
APOSPC6759-25G
4-Fluor-L-phenylalanin 98%
4-Fluor-L-phenylalanin
Formel:
C₉H₁₀FNO₂ Molecular Weight: 183,2 g/mol Schmelzpunkt: 247…249 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00063064 CAS-Nummer: 1132-68-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...