Tridecafluoroheptanamide

Lieferant: Apollo Scientific

Synonyme: Perfluoroheptanamide

PC6768-25G PC6768-5G
APOSPC6768-25GEA 0 EUR
APOSPC6768-25G APOSPC6768-5G
Tridecafluoroheptanamide
Tridecafluoroheptanamide
Formel: C₇H₂F₁₃NO
Molecular Weight: 363.08 g/mol
Schmelzpunkt: 121.5…122.5 °C
Flammpunkt: >110 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00407510
CAS-Nummer: 2358-22-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR