1,1,2,2-Tetrafluor-1-iodethan 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 1,1,2,2-Tetrafluorethyliodid

PC6783-1G PC6783-5G
APOSPC6783-1GEA 206 EUR
APOSPC6783-1G APOSPC6783-5G
1,1,2,2-Tetrafluor-1-iodethan 97%
1,1,2,2-Tetrafluor-1-iodethan
Formel: C₂HF₄I
Molecular Weight: 227.93 g/mol
Siedepunkt: 41…42 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00069111
CAS-Nummer: 354-41-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR