Tetrafluorhydrochinon 95%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
2,3,5,6-Tetrafluorohydroquinone, Tetrafluorobenzene-1,4-diol
PC6790-1G
PC6790-25G
PC6790-5G
APOSPC6790-1GEA
43.5
EUR
APOSPC6790-1G
APOSPC6790-25G
APOSPC6790-5G
Tetrafluorhydrochinon 95%
Tetrafluorhydrochinon
Formel:
C₆H₂F₄O₂ Molecular Weight: 182.07 g/mol Schmelzpunkt: 168…174 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00002340 CAS-Nummer: 771-63-1 EINECS: 212-237-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...