Tetrafluorisophthalonitril 98%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
Tetrafluorbenzol-1,3-dicarbonitril, Tetrafluorisophthalsäuredinitril
PC6797-1G
PC6797-25G
PC6797-5G
APOSPC6797-1GEA
21.9
EUR
APOSPC6797-1G
APOSPC6797-25G
APOSPC6797-5G
Tetrafluorisophthalonitril 98%
Tetrafluorisophthalonitril
Formel:
C₈F₄N₂ Molecular Weight: 200,1 g/mol Schmelzpunkt: 76…79 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00013290 CAS-Nummer: 2377-81-3 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...