2,2,3,3-Tetrafluoropropionic acid 97%
Lieferant: Apollo Scientific
|
Gefahr
|
PC6866E-25G
PC6866E-5G
APOSPC6866E-25GEA
183
EUR
APOSPC6866E-25G
APOSPC6866E-5G
2,2,3,3-Tetrafluoropropionic acid 97%
2,2,3,3-Tetrafluoropropionic acid
Formel:
C₃H₂F₄O₂ Molecular Weight: 146,04 g/mol Siedepunkt: 133…134 °C Dichte: 1,563 g/cm³ (25 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00054686 CAS-Nummer: 756-09-2 EINECS: 212-049-7 UN: 3265 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...