2-Bromo-3-fluorobenzyl bromide 99%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
2-Bromo-1-(bromomethyl)-3-fluorobenzene
PC6878-1G
PC6878-25G
PC6878-5G
APOSPC6878-1GEA
30.6
EUR
APOSPC6878-1G
APOSPC6878-25G
APOSPC6878-5G
2-Bromo-3-fluorobenzyl bromide 99%
2-Bromo-3-fluorobenzyl bromide
Formel:
C₇H₅Br₂F Molecular Weight: 267,923 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD12911576 CAS-Nummer: 1184918-22-6 UN: 3261 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...