Methyl-3-oxo-3-[6-(trifluoromethyl)pyridin-3-yl]propanoate 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: Methyl 3-oxo-3-(6-(trifluoromethyl)pyridin-3-yl)propanoate

PC6886-1G PC6886-5G
APOSPC6886-1GEA 203 EUR
APOSPC6886-1G APOSPC6886-5G
Methyl-3-oxo-3-[6-(trifluoromethyl)pyridin-3-yl]propanoate 95%
Methyl-3-oxo-3-[6-(trifluoromethyl)pyridin-3-yl]propanoate
Formel: C₁₀H₈F₃NO₃
Molecular Weight: 247.18 g/mol
Schmelzpunkt: 60…62 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01862654
CAS-Nummer: 386704-15-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR