4-Fluor-3-formylphenylboronsäure 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-Fluoro-3-formylbenzeneboronic acid, (4-Fluoro-3-formylphenyl)boronic acid
PC6909-25G
PC6909-5G
APOSPC6909-25GEA
55.1
EUR
APOSPC6909-25G
APOSPC6909-5G
4-Fluor-3-formylphenylboronsäure 98%
4-Fluor-3-formylphenylboronsäure
Formel:
C₇H₆BFO₃ Molecular Weight: 167,93 g/mol Schmelzpunkt: 215…220 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD02093074 CAS-Nummer: 374538-01-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...