5-Fluor-2-hydroxypyridin

Lieferant: Apollo Scientific
Achtung

PC6957-1G PC6957-5G PC6957-10G
APOSPC6957-1GEA 21.9 EUR
APOSPC6957-1G APOSPC6957-5G APOSPC6957-10G
5-Fluor-2-hydroxypyridin
5-Fluor-2-hydroxypyridin
Formel: C₅H₄FNO
Molecular Weight: 113.09 g/mol
Schmelzpunkt: 152…155 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03092918
CAS-Nummer: 51173-05-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR