4-(2-Fluorphenylcarbamoyl)phenylboronsäure 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: (4-((2-Fluorophenyl)carbamoyl)phenyl)boronic acid

PC6986-5G
APOSPC6986-5GEA 108 EUR
APOSPC6986-5G
4-(2-Fluorphenylcarbamoyl)phenylboronsäure 98%
4-(2-Fluorphenylcarbamoyl)phenylboronsäure
Formel: C₁₃H₁₁BFNO₃
Molecular Weight: 259,0408 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD09027205
CAS-Nummer: 874288-06-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR