3-Chlor-2,6-difluorphenylboronsäure 98%
Lieferant: Apollo Scientific
Synonyme:
(3-Chloro-2,6-difluorophenyl)boronic acid
PC7011-1G
PC7011-5G
APOSPC7011-1GEA
69.6
EUR
APOSPC7011-1G
APOSPC7011-5G
3-Chlor-2,6-difluorphenylboronsäure 98%
3-Chlor-2,6-difluorphenylboronsäure
Formel:
C₆H₄BClF₂O₂ Molecular Weight: 192.3556 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD04038753 CAS-Nummer: 1031226-45-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...