3-Fluorphenylboronsäure 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(3-Fluorphenyl)metaborsäure, 3-Fluorbenzolboronsäure, Dihydroxy(3-fluorphenyl)boran
PC7016-25G
PC7016-5G
PC7016-100G
APOSPC7016-25GEA
42.2
EUR
APOSPC7016-25G
APOSPC7016-5G
APOSPC7016-100G
3-Fluorphenylboronsäure 97%
3-Fluorphenylboronsäure
Formel:
C₆H₆BFO₂ Molecular Weight: 139,92 g/mol Schmelzpunkt: 211…221 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00236042 CAS-Nummer: 768-35-4 EINECS: 476-720-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...