2-Cyan-4-fluorphenylboronsäurepinakolester
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Cyano-4-fluorophenylboronic Acid Pinacol Ester, 2-Cyano-4-fluorobenzeneboronic acid pinacol ester
PC7020-5G
APOSPC7020-5GEA
168
EUR
APOSPC7020-5G
2-Cyan-4-fluorphenylboronsäurepinakolester
2-Cyan-4-fluorphenylboronsäurepinakolester
Formel:
C₁₃H₁₅BFNO₂ Molecular Weight: 247,07 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09260444 CAS-Nummer: 461451-63-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...