2,3-Difluor-6-methoxyphenylboronsäure 98%
Lieferant: Apollo Scientific
|
Achtung
|
PC7025-1G
PC7025-5G
PC7025-25G
APOSPC7025-1GEA
130
EUR
APOSPC7025-1G
APOSPC7025-5G
APOSPC7025-25G
2,3-Difluor-6-methoxyphenylboronsäure 98%
2,3-Difluor-6-methoxyphenylboronsäure
Formel:
C₇H₇BF₂O₃ Molecular Weight: 187,94 g/mol Schmelzpunkt: 125…130 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD03095116 CAS-Nummer: 957061-21-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...