3,4-Difluor-2-methylphenylboronsäurepinakolester 95%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-(3,4-Difluoro-2-methylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
PC7031-1G
PC7031-250MG
APOSPC7031-1GEA
82.8
EUR
APOSPC7031-1G
APOSPC7031-250MG
3,4-Difluor-2-methylphenylboronsäurepinakolester 95%
3,4-Difluor-2-methylphenylboronsäurepinakolester
Formel:
C₁₃H₁₇BF₂O₂ Molecular Weight: 254.08 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD08669570 CAS-Nummer: 1025707-97-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...