2,6-Difluor-4-pyridinboronsäure

Lieferant: Apollo Scientific
Achtung

Synonyme: (2,6-Difluoropyridin-4-yl)boronic acid, 2,6-Difluoro-4-pyridylboronic acid

PC7034-100MG PC7034-250MG
APOSPC7034-100MGEA 45.1 EUR
APOSPC7034-100MG APOSPC7034-250MG
2,6-Difluor-4-pyridinboronsäure
2,6-Difluoro-4-pyridineboronic acid
Formel: C₅H₄BF₂NO₂
Molecular Weight: 158.8986 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD04114289
CAS-Nummer: 401816-16-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR