3-Fluor-2-methoxyphenylboronsäure

Lieferant: Apollo Scientific
Achtung

PC7044-1G PC7044-5G
APOSPC7044-1GEA 33.5 EUR
APOSPC7044-1G APOSPC7044-5G
3-Fluor-2-methoxyphenylboronsäure
3-Fluor-2-methoxyphenylboronsäure
Formel: C₇H₈BFO₃
Molecular Weight: 169.9460 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD03095355
CAS-Nummer: 762287-59-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR