4'-Chlor-2'-fluor-5'-methylacetophenon
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1-(4-Chloro-2-fluoro-5-methylphenyl)ethanone
PC7084-1G
PC7084-250MG
APOSPC7084-1GEA
56.8
EUR
APOSPC7084-1G
APOSPC7084-250MG
4'-Chlor-2'-fluor-5'-methylacetophenon
4'-Chlor-2'-fluor-5'-methylacetophenon
Formel:
C₉H₈ClFO Molecular Weight: 186,6106 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00236601 CAS-Nummer: 177211-26-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...