7-Bromo-5-fluoroindole 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 7-Bromo-5-fluoro-1H-indole

PC7087-1G PC7087-250MG
APOSPC7087-1GEA 198 EUR
APOSPC7087-1G APOSPC7087-250MG
7-Bromo-5-fluoroindole 97%
7-Bromo-5-fluoroindole
Formel: C₈H₅BrFN
Molecular Weight: 214,04 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD04037875
CAS-Nummer: 408355-23-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR